Learn More
Bon Opus Biosciences Recombinant Human Complement Factor H-related Protein 2/CFHR2 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP16350UG
Encompass_Preferred
Recombinant Human Complement Factor H-Related 2 is produced by our Mammalian expression system and the target gene encoding Glu19-Lys270 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P36980. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# BP163
Catalog No. 50-208-0259