Learn More
Bon Opus Biosciences Recombinant Human Complement Factor H-related 5/CFHR5 (C-6His) 500ug
Supplier: Bon Opus Biosciences CC74500UG
Recombinant Human CFHR5 is produced by our Mammalian expression system and the target gene encoding Glu19-Glu569 is expressed with a 6His tag at the C-terminus. Accession# Q9BXR6. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CC74