Learn More
Bon Opus Biosciences Recombinant Human CGA/CGB3 (C-Flag,C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CB8450UG
Encompass_Preferred
Recombinant Human Glycoprotein hormones alpha chain / Choriogonadotropin subunit beta 3 is produced by our Mammalian expression system and the target gene encoding Ala25-Ser116/Ser21-Gln165 is expressed with a Flag tag at the C-terminus, 6His tag at the C-terminus. Expression system: Human Cells. Accession# P01215/P0DN86. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CB84
Catalog No. 50-208-0377