Learn More
Bon Opus Biosciences Recombinant Human Neuronal Acetylcholine Receptor Subunit β-3/CHRNB3 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CI32500UG
Encompass_Preferred
Recombinant Human CHRNB3 is produced by our Mammalian expression system and the target gene encoding Ile25-Leu232 is expressed with a 6His tag at the C-terminus. Accession# Q05901. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI32
Catalog No. 50-217-2801