Learn More
Bon Opus Biosciences Recombinant Human Ciliary Neurotrophic Factor/CNTF -500ug
SDP
Supplier:  Bon Opus Biosciences BP084500UG
Encompass_Preferred
Recombinant Human Ciliary Neurotrophic Factor is produced by E.coli. The target gene encoding A2-P187 is expressed. Expression system: E. coli. Accession# P26441. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM Tris, 30umM NaCl, 0.5%CHAPS, 0.umM TCEP, 0.umM EDTA, pH8.0. Bon Opus Cat# BP084
Catalog No. 50-208-0159