Learn More
BioVendor, LLC Human CXCL12 (SDF-1) (Biotinylated) E.coli Recombinant Protein (0.01 mg)
SDP
Supplier:  BioVendor, LLC RD172381010BIOTIN
Encompass_Preferred
Human CXCL12 (SDF-1) (Biotinylated) E.coli Recombinant Protein (0.01 mg). DATA: Total 87 AA. MW: 10,4 kDa. Biotinylated at the last Lysine in sequence. UniProtKB acc# P48061 2 (2489) Protein ID confirmed by LC-MS/MS. Purity by SDS PAGE: >97 %. BIO ACTIVITY: EC50 = 2.5nM determined by Calcium Flux with human CXCR4 in U937 cells. Migration confirmed with U937 cells expressing CXCR4. ENDOTOXIN LVL: <0.01 EU/ug. APPLICATIONS: Western blot, Immunocytochemistry, ELISA, Cell culture, animal studies, Biologically active, Flow cytometry. OTHER NAMES: SDF-1alpha, SDF1A, SDF 1A, Stromal cell-derived factor 1, SDF-1a, C-X-C motif chemokine 12, Pre-B cell growth-stimulating factor, PBSF, hIRH, CXCL12 (isoform alpha) SUMMARY: CXCL12 is an important factor in physiological and pathological processes, including embryogenesis, hematopoiesis, angiogenesis and inflammation, as it activates and/or induces migration of hematopoietic progenitor and stem cells, endothelial cells and most leukocytes.
Catalog No. 50-196-3869
 Restricted Item