Learn More
Bon Opus Biosciences Recombinant Human CXCL6 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C59850UG
Encompass_Preferred
Recombinant Human C-X-C Motif Chemokine 6 is produced by our Mammalian expression system and the target gene encoding Gly38-Asn114 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# P80162. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 5% Trehalose, pH 7.4. Bon Opus Cat# C598
Catalog No. 50-208-0357