Learn More
Bon Opus Biosciences Recombinant Human D-Tyrosyl-tRNA(Tyr) Deacylase 1/DTD1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CI721MG
Encompass_Preferred
Recombinant Human DTD1 is produced by our Mammalian expression system and the target gene encoding Met1-Pro209 is expressed with a 6His tag at the C-terminus. Accession# Q8TEA8. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI72
Catalog No. 50-217-2803