Learn More
Bon Opus Biosciences Recombinant Human DCN1-Like Protein 1/DCUN1D1 (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C271500UG
Encompass_Preferred
Recombinant Human DCN1-Like Protein 1 is produced by our E.coli expression system and the target gene encoding Met1-Val259 is expressed with a 6His tag at the N-terminus. Accession# Q96GG9. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C271
Catalog No. 50-217-2732