Learn More
Bon Opus Biosciences Recombinant Human Beta-Defensin 4/DEFB4 -50ug
SDP
Supplier:  Bon Opus Biosciences BP02350UG
Encompass_Preferred
Recombinant Human beta-Defensin 4 is produced by our E.coli expression system and the target gene encoding Glu23-Pro72 is expressed. Expression system: E. coli. Accession# Q8WTQ1. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 130mM NaCl, pH 7.4. Bon Opus Cat# BP023
Catalog No. 50-208-0104