Learn More
Lonza Walkersville Hu Dental Pulp SC Basal Medium
Catalog No. 5000496282
Encompass
The DPSC Dental Pulp Stem Cell BulletKit Medium is offered as a BulletKit Medium (catalog no PT-3005) which includes both the basal media and the necessary supplements for proliferation of human dental pulp derived mesenchymal stem cells The basal medium (DPSC Basal Medium catalog no PT-3927) and the necessary supplements (DPSC SingleQuot Kit catalog no PT-4516) can also be purchased individually
Catalog No. 50-004-96282 Supplier Lonza Walkersville Supplier No. PT3927
The DPSC Dental Pulp Stem Cell BulletKit Medium is offered as a BulletKit Medium (catalog no PT-3005) which includes both the basal media and the necessary supplements for proliferation of human dental pulp derived mesenchymal stem cells The basal medium (DPSC Basal Medium catalog no PT-3927) and the necessary supplements (DPSC SingleQuot Kit catalog no PT-4516) can also be purchased individually