Learn More
Bon Opus Biosciences Recombinant Human EIF1A, X-Chromosomal/EIF1AX (N-6His) 1mg
Supplier: Bon Opus Biosciences C2191MG
Recombinant Human EIF1AX is produced by our E.coli expression system and the target gene encoding Met1-Ile144 is expressed with a 6His tag at the N-terminus. Accession# P47813. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# C219