Learn More
Bon Opus Biosciences Recombinant Human Ephrin A Receptor 2/EphA2 (C-Fc) 500ug
SDP
Supplier:  Bon Opus Biosciences CP37500UG
Encompass_Preferred
Recombinant Human Ephrin A Receptor 2 is produced by our Mammalian expression system and the target gene encoding Ala24-Asn534 is expressed with a Fc tag at the C-terminus. Accession# P29317. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CP37
Catalog No. 50-217-2824