Learn More
Bon Opus Biosciences Recombinant Human EPO-R (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW0650UG
Encompass_Preferred
Recombinant Human Erythropoietin Receptor is produced by our Mammalian expression system and the target gene encoding Ala25-Pro250 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# P19235. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW06
Catalog No. 50-208-0409