Learn More
Bon Opus Biosciences Recombinant Human Endoplasmic Reticulum Resident Protein 27/ERP27 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CA661MG
Encompass_Preferred
Recombinant Human ERP27 is produced by our Mammalian expression system and the target gene encoding Glu26-Leu273 is expressed with a 6His tag at the C-terminus. Accession# Q96DN0. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CA66
Catalog No. 50-217-2771