Learn More
Bon Opus Biosciences Recombinant Human FAM172A (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP010500UG
Encompass_Preferred
Recombinant Human FAM172A is produced by Human 293 Cells. The target gene encoding Gln19-Lys416 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# Q8WUF8. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PB, 15umMNacl pH 7.4. Bon Opus Cat# BP010
Catalog No. 50-208-0091