Learn More
Bon Opus Biosciences Recombinant Human Fc γ RIIb/CD32b (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences BP1311MG
Encompass_Preferred
Recombinant Human Fc-gamma RIIb is produced by our Mammalian expression system and the target gene encoding Thr43-Pro217 is expressed with a 6His tag at the C-terminus. Accession# P31994. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C444
Catalog No. 50-217-2743