Learn More
Bon Opus Biosciences Recombinant Human FGF-4 -50ug
SDP
Catalog No. 502080401
Encompass_Preferred
Recombinant Human Fibroblast Growth Factor 4 is produced by our E.coli expression system and the target gene encoding Ser71­-Leu206 is expressed. Expression system: E.coli. Accession# P08620. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR68
Catalog No. 50-208-0401 Supplier Bon Opus Biosciences Supplier No. CR6850UG
Recombinant Human Fibroblast Growth Factor 4 is produced by our E.coli expression system and the target gene encoding Ser71­-Leu206 is expressed. Expression system: E.coli. Accession# P08620. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR68