Learn More
Bon Opus Biosciences Recombinant Human FGF R5/FGFRL1 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C520500UG
Encompass_Preferred
Recombinant Human FGFRL1 is produced by our Mammalian expression system and the target gene encoding Ala25-Pro378 is expressed with a 6His tag at the C-terminus. Accession# Q8N441. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Bon Opus Cat# C520
Catalog No. 50-217-2749