Learn More
Bon Opus Biosciences Recombinant Human FGL1 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW5550UG
Encompass_Preferred
Recombinant Human Fibrinogen-like Protein 1 is produced by our Mammalian expression system and the target gene encoding Leu23-Ile312 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q08830. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris,300mM NaCl, 10% Glycerol,2mMEDTA,pH 8.0. Bon Opus Cat# CW55
Catalog No. 50-208-0437