Learn More
Bon Opus Biosciences Recombinant Human FGL1 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW6850UG
Encompass_Preferred
Recombinant Human Fibrinogen-like Protein 1 is produced by our Mammalian expression system and the target gene encoding Leu23-Ile312 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q08830. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% trehalose, pH7.4. Bon Opus Cat# CW68
Catalog No. 50-208-0449