Learn More
Bon Opus Biosciences Recombinant Human Fascin/FSCN1 -500ug
SDP
Supplier:  Bon Opus Biosciences BP179500UG
Encompass_Preferred
Recombinant Human Fascin is produced by our E.coli expression system and the target gene encoding Met1-Tyr493 is expressed. Expression system: E. coli. Accession# Q16658. Formulation: Supplied as a 0.2 um filtered solution of 25mM Tris,100mM NaCl,2mM DTT,10% Glycerol, pH8.0. Bon Opus Cat# BP179
Catalog No. 50-208-0286