Learn More
Bon Opus Biosciences Recombinant Human Follitropin Subunit Beta/FSHB (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CA131MG
Encompass_Preferred
Recombinant Human FSHB is produced by our Mammalian expression system and the target gene encoding Asn19-Glu129 is expressed with a 6His tag at the C-terminus. Accession# P01225. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4 Bon Opus Cat# CA13
Catalog No. 50-217-2767