Learn More
Bon Opus Biosciences Recombinant Human Ferritin heavy chain/FTH -500ug
SDP
Supplier:  Bon Opus Biosciences BP159500UG
Encompass_Preferred
Recombinant Human Ferritin heavy chain is produced by our E.coli expression system and the target gene encoding Met1-Ser183 is expressed. Expression system: E. coli. Accession# P02794. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP159
Catalog No. 50-208-0252