Learn More
Bon Opus Biosciences Recombinant Human Ferritin Light Chain/FTL (N-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP16050UG
Encompass_Preferred
Recombinant Human Ferritin light chain is produced by our E.coli expression system and the target gene encoding Met1-Asp175 is expressed with a 6His tag at the N-terminus. Expression system: E. coli. Accession# P02792. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM Tris, 250mM NaCl, 1mM EDTA, pH9.5. Bon Opus Cat# BP160
Catalog No. 50-208-0253