Learn More
Bon Opus Biosciences Recombinant Human GADD45β/GADD45B (N-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CE371MG
Encompass_Preferred
Recombinant Human GADD45B is produced by our E.coli expression system and the target gene encoding Met1-Arg160 is expressed with a 6His tag at the N-terminus. Accession# O75293. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CE37
Catalog No. 50-217-2777