Learn More
Bon Opus Biosciences Recombinant Human GAS6 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C01W50UG
Encompass_Preferred
Recombinant Human Growth Arrest-specific Protein 6 is produced by our Mammalian expression system and the target gene encoding Ala31-Ala678 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q14393. Formulation: Supplied as a 0.2 um filtered solution of PBS, 10% Glycerol , pH7.4. Bon Opus Cat# C01W
Catalog No. 50-208-0337