Learn More
Bon Opus Biosciences Recombinant Human β-Galactosidase/GLB1 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C473500UG
Encompass_Preferred
Recombinant Human beta-Galactosidase is produced by our Mammalian expression system and the target gene encoding Leu24-Val677 is expressed with a 6His tag at the C-terminus. Accession# P16278. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Bon Opus Cat# C473
Catalog No. 50-217-2746