Learn More
Bon Opus Biosciences Recombinant Human beta-Galactosidase/GLB1(C-His) - Source: Human 293 Cells
SDP
Catalog No. NC2733621
Encompass_Preferred
Recombinant Human beta-Galactosidase is produced by our Mammalian expression system and the target gene encoding Leu24-Val677 is expressed with a 6His tag at the C-terminus. Accession#P16278. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM Tris-HCl, 150mM NaCl, pH 8.0
Catalog No. NC2733621 Supplier Bon Opus Biosciences Supplier No. BP14750UG
Recombinant Human beta-Galactosidase is produced by our Mammalian expression system and the target gene encoding Leu24-Val677 is expressed with a 6His tag at the C-terminus. Accession#P16278. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM Tris-HCl, 150mM NaCl, pH 8.0