Learn More
Cell Systems Corporation Primary Human Glomerular Mesangial Cells (ACBRI 127 V)
SDP
Supplier:  Cell Systems Corporation ACBRI 127 V
Encompass_Preferred
Primary Human Glomerular Mesangial Cells (ACBRI 127 V) were initiated by elutriation of dispase dissociated decapsulated glomeruli isolated from normal human cortical kidney tissue. Unlike cells from other providers, our cells are isolated without antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.
Catalog No. NC0824568