Learn More
Bon Opus Biosciences Recombinant Human Glypican-1/GPC1 (N-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP17350UG
Encompass_Preferred
Recombinant Human Glypican-1 is produced by our Mammalian expression system and the target gene encoding Asp24-Thr529 is expressed fused with a 6His tag at the N-terminus. Expression system: Human 293 Cells. Accession# P35052. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP173
Catalog No. 50-208-0275