Learn More
Bon Opus Biosciences Recombinant Human Heat Shock Protein β-8//HSPB8 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CH75500UG
Encompass_Preferred
Recombinant Human Heat shock protein beta-8 is produced by our E.coli expression system and the target gene encoding Met1-Thr196 is expressed with a 6His tag at the C-terminus. Accession# Q9UJY1. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CH75
Catalog No. 50-217-2795