Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HIF1A ELISA Kit

Catalog No. enehif1a
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENEHIF1A 96 Tests
ENEHIF1A2 2 x 96 Tests
ENEHIF1A5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ENEHIF1A Supplier Invitrogen™ Supplier No. EHIF1A
Only null left
Add to Cart
Edge
Add to Cart

ELISA

The Human HIF1A ELISA quantitates Hu HIF1A in human serum, plasma, or cell culture lysates. The assay will exclusively recognize both natural and recombinant Hu HIF1A. Principle of the method The Human HIF1A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16665
Assay Range 81.92 to 20,000 pg/mL
Assay Sensitivity 30 pg/mL
Description HIF-1 Alpha ELISA Kit, 96 Assays, 1 Kit, Human Species, Immunology Research Category, Colorimetric Detection Method, Colorimetric Elisa (TMB Substrate) in Clear 96-Well Strip Plate, Contain Precoated 96-Well Strip Plate(S) and Lot-Optimized Reagents
Assay 96
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Protein Family Transcription factors
Sample Type Cell Lysates, Plasma, Serum
Specificity Specific to human HIF-1 A and shows cross-reactivity with mouse and rat HIF-1 A
Substrate Type TMB
Sufficient For 96 Assays
Target Species Rat, Mouse
For Use With (Application) Protein Assays and Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3091
Gene Symbol HIF-1-alpha,HIF-1 A,HIF-1 alpha,HIF1,HIF1-ALPHA,MOP1,PASD8,bHLHe78
Host Species Human
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Measurement Absorbance at 450 nm minus absorbance (at 550 nm)
Measuring Range 450 to 550 nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Immunology
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL
Storage Requirements 2°C to 8°C
Target HIF1A
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.