Learn More
Bon Opus Biosciences Recombinant Human Heat Shock Protein β-2/HSPB2/MKBP (C-6His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP021500UG
Encompass_Preferred
Recombinant Human Heat shock protein beta-2 is produced by our E.coli expression system and the target gene encoding Met1-Pro182 is expressed with a 6His tag at the C-terminus. Expression system: E. coli. Accession# Q16082. Formulation: Supplied as a 0.2 um filtered solution of 20mM HEPES, 0.1M KCl, pH 7.5. Bon Opus Cat# BP021
Catalog No. 50-208-0101