Learn More
Bon Opus Biosciences Recombinant Human IL-2 Receptor Subunit β/IL-2RB/CD122 (C-6His) 500UG
SDP
Supplier:  Bon Opus Biosciences CJ81500UG
Encompass_Preferred
Recombinant Human IL-2 receptor beta is produced by our Mammalian expression system and the target gene encoding Ala27-Asp239 is expressed with a 6His tag at the C-terminus. Accession# P14784. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,pH7.4. Bon Opus Cat# CJ81
Catalog No. 50-217-2815