Learn More
Bon Opus Biosciences Recombinant Human Interferon gamma/IFN-gamma (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP069500UG
Encompass_Preferred
Recombinant Human Interferon gamma is produced by Human 293 Cells. The target gene encoding Q24-G161 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P01579. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP069
Catalog No. 50-208-0144