Learn More
Bon Opus Biosciences Recombinant Human Interferon α/β Receptor 2/IFNAR2 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C4761MG
Encompass_Preferred
Recombinant Human Interferon alpha/beta Receptor 2 is produced by our Mammalian expression system and the target gene encoding Ile27-Lys243 is expressed with a 6His tag at the C-terminus. Accession# P48551. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C476
Catalog No. 50-217-2747