Learn More
Bon Opus Biosciences Recombinant Human Insulin-Like Growth Factor I/IGF-I/IGF1 (4-70) -50ug
SDP
Catalog No. 502080350
Encompass_Preferred
Recombinant Human Insulin-like Growth Factor I is produced by our E.coli expression system and the target gene encoding Thr52-Ala118 is expressed. Expression system: E. coli. Accession# P05019. Formulation: Lyophilized from a 0.2 μm filtered solution of 300mM NaAc, pH 6.5. Bon Opus Cat# C031
Catalog No. 50-208-0350 Supplier Bon Opus Biosciences Supplier No. C03150UG
Recombinant Human Insulin-like Growth Factor I is produced by our E.coli expression system and the target gene encoding Thr52-Ala118 is expressed. Expression system: E. coli. Accession# P05019. Formulation: Lyophilized from a 0.2 μm filtered solution of 300mM NaAc, pH 6.5. Bon Opus Cat# C031