Learn More
Bon Opus Biosciences Recombinant Human IGF binding Protein 7/IGFBP-7 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences BP209500UG
Encompass_Preferred
Recombinant Human Insulin-like Growth Factor-binding Protein 7 is produced by our Mammalian expression system and the target gene encoding Ser27-Leu282 is expressed with a 6His tag at the C-terminus. Accession# Q16270. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,pH7.4. Bon Opus Cat# CU50
Catalog No. 50-217-2831