Learn More
Bon Opus Biosciences Recombinant Human IgG3 Fc -50ug
SDP
Supplier:  Bon Opus Biosciences FC0950UG
Encompass_Preferred
Recombinant Human Immunoglobulin Heavy Constant Gamma 3 is produced by our Mammalian expression system and the target gene encoding Glu99-Lys377 is expressed. Expression system: Human Cells. Accession# P01860. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# FC09
Catalog No. 50-208-0576