Learn More
Bon Opus Biosciences Recombinant Human Interleukin-12 Subunit Beta/IL-12B (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP088500UG
Encompass_Preferred
Recombinant Human Interleukin-12 Subunit Beta is produced by Human 293 Cells. The target gene encoding I23-S328 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# P29460. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP088
Catalog No. 50-208-0163