Learn More
Bon Opus Biosciences Recombinant Human Interleukin-1β/IL-1β 1MG
SDP
Supplier:  Bon Opus Biosciences CG931MG
Encompass_Preferred
Recombinant Human Interleukin-1 beta is produced by our E.coli expression system and the target gene encoding Ala117-Ser269 is expressed. Accession# P01584. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CG93
Catalog No. 50-217-2790