Learn More
Bon Opus Biosciences Recombinant Human Interleukin-22/IL-22 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP087500UG
Encompass_Preferred
Recombinant Human Interleukin-22 is produced by Human 293 Cells. The target gene encoding A34-I179 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# Q9GZX6. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP087
Catalog No. 50-208-0162