Learn More
Bon Opus Biosciences Recombinant Human IL-22(N297G) (C-hIgG1 Fc) 50ug
Supplier: Bon Opus Biosciences BP108150UG
Recombinant Human IL-22(N297G) (C-hIgG1 Fc) is produced by Human Cells. The target gene encoding Ala34-Ile 179 is expressed with a C-hIgG1 Fc tag. Accession# Q9GZX6-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1081