Learn More
Bon Opus Biosciences Recombinant Human IL-23 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CW54500UG
Encompass_Preferred
Recombinant Human Interleukin-23 is produced by our Mammalian expression system and the targetgene encoding Ile23-Ser328&Arg20-Pro189 is expressed with a 6His tag at the C-terminus. Accession# P29460&Q9NPF7. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4 Bon Opus Cat# CW54
Catalog No. 50-217-2834