Learn More
Bon Opus Biosciences Recombinant Human IL-23 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW9350UG
Encompass_Preferred
Recombinant Human Interleukin-23  is produced by our Mammalian expression system and the target gene encoding Ile23-Ser328/Ala21-Pro189 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# P29460/Q9NPF7. Formulation: Lyophilized from a 0.2 um filtered solution of 20mMPB,150mMNaCl,pH7.4. Bon Opus Cat# CW93
Catalog No. 50-208-0470