Learn More
Bon Opus Biosciences Recombinant Human IL-23A / mouse IL-12B Heterodimer (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW8150UG
Encompass_Preferred
Recombinant Chimeric Human/Mouse Interleukin-23  is produced by our Mammalian expression system and the target gene encoding Arg20-Pro189/Met23-Ser335 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q9NPF7/P43432. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW81
Catalog No. 50-208-0461