Learn More
Bon Opus Biosciences Biotinylated Recombinant Human Interleukin-33/IL-33 -25ug
SDP
Supplier:  Bon Opus Biosciences BP18425UG
Encompass_Preferred
Biotinylated recombinant Human IL-33 is produced by E.coli. The target gene encoding S111-T270 is expressed with a N-AVI tag. Expression system: E. coli. Accession# O95760. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP184
Catalog No. 50-208-0293