Learn More
Bon Opus Biosciences Recombinant Human IL-36 Beta(153AA) -50ug
SDP
Supplier:  Bon Opus Biosciences CR5950UG
Encompass_Preferred
Recombinant Human Interleukin-36 Beta/Il36B is produced by our E.coli expression system and the target gene encoding Arg5-Glu157 is expressed. Expression system: E.coli. Accession# Q9NZH7-2. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CR59
Catalog No. 50-208-0392