Learn More
Bon Opus Biosciences Recombinant Human IL-36 Beta(157AA) -50ug
SDP
Supplier:  Bon Opus Biosciences CR5750UG
Encompass_Preferred
Recombinant Human IL-36 Beta is produced by our E.coli expression system and the target gene encoding Met1-Glu157 is expressed. Expression system: E.coli. Accession# Q9NZH7-2. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR57
Catalog No. 50-208-0390